Yeast Extract, Powder, Certified, HiMedia Laboratories
Supplier: HiMedia
CR027-500G
CR027-25KG
95041-228EA
203.38
USD
95041-228
95000-018
Yeast Extract, Powder, Certified, HiMedia Laboratories
Media for Microbiology
Dehydrated Media for Microbiology
It is a certified grade manufactured under controlled condition by autolysis of Saccharomyces cerevesiae
- Light yellow to brownish yellow colored
- Homogenous, free flowing powder with characteristic odour but not putrescent
- Soluble in distilled/purified water
- Insoluble in alcohol and ether
- Reaction of 2% w/v aqueous solution at 25 ºC
It is rich in vitamins, especially those belonging to B-complex and used particularly for cultivation of microorganisms encountered in milk or other dairy products.
Caution: Store below 30 °C.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...