Tryptone Phosphate Broth, HiMedia Laboratories
Supplier: HiMedia
M953-500G M953-2.5KG M953-5KG
95022-526EA 305.95 USD
95022-526 89226-938 89226-940
Tryptone Phosphate Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Tryptone phosphate broth is used for enrichment and cultivation of enteropathogenic Escherichia coli from suspected food samples.
  • Cream to yellow colored
  • Homogeneous, free flowing powder
  • Reaction of 3.05% w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and the prepared medium at 2 to 8 °C.