Deoxynivalenol ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
BTDOEK-001
103388-984EA 523.08 USD
103388-984
Deoxynivalenol ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Deoxynivalenol ELISA , allows to detect the Deoxynivalenol in food and feed crop (cereals).
  • Dynamic Range: 3.1-120ng/ml
  • Sensitivity: 12ppb
  • Incubation: 0.75 hours
Deoxynivalenol (DON), also known as Vomitoxin, is a type of mycotoxin (toxic mould fungus). Chemically it belongs to type B trichothecenes. The mycotoxin DON is formed as a poisonous metabolite from the Fusarium genus, i.e. F. culmorum and F. graminearum. DON enters into the human food chain via contamination of plant-based foods. It is abundant in grains such as wheat, barley, oats, rye, maize and processed grains such as malt, beer and bread. The main toxin production by Fusarium species happens in the field. Climate conditions have a big influence on contamination.The harvest and storage conditions play also an important role in DON contamination. DON is a potent inhibitor of protein synthesis and thus acts generally cause cell damage.The mycotoxin DON has been connected with incidents of mycotoxicoses in both humans and farm animals.