Anti-Aprotinin Antibodies (IgG) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
APR31-K01
102993-928EA 1516.92 USD
102993-928
Anti-Aprotinin Antibodies (IgG) ELISA, Eagle Biosciences
Assays ELISAs
Anti-Aprotinin IgG Antibodies ELISA is designed to detect IgG Aprotinin Antibodies in serum samples.
  • Dynamic Range: 12.5 U/ml - 200 U/ml
  • Sensitivity: 12.5 U/ml
  • Incubation: 2 hours
The Eagle Biosciences Anti-Aprotinin IgG Antibodies ELISA Assay Kit is designed for the quantitative determination of IgG Aprotinin Antibodies in serum samples. The Anti-Aprotinin Antibodies ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.