Folic Acid ELISA, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
FOL06-K01
102994-396EA 1124.62 USD
102994-396
Folic Acid ELISA, Eagle Biosciences, Inc.
Assays ELISAs
Folic Acid ELISA is intended to detect Folic Acid in food products by enzyme linked immuno ELISA.
  • Dynamic Range: 4 - 400 ng/ml
  • Sensitivity: 2 ng/ml
  • Incubation: 2 hours
Folates play an important role in the synthesis of nucleic acids and some amino acids and gained re­cently increased interest because they belong to the group of antioxidative vitamins. In the last years the in­fluence of folic acid supplementation to avoid abortion and dysraphism was a topic of research increasingly. Folic acid as the most stable representative of the group of folates is added to a broad range of food.