Human CD44 / Pgp-1 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C4431-K01
102994-056EA 830.38 USD
102994-056
Human CD44 / Pgp-1 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD44 ELISA detects CD44 in human serum, plasma, buffered solutions or cell culture medium.
  • Dynamic Range: 0.31-10 ng/ml
  • Sensitivity: 11 ng/ml
  • Incubation: 2h10
The Eagle Biosciences CD44 ELISA is to be used for the in-vitro quantitative determination of soluble CD44 (CD44/phagocytic glycoprotein-1/Pgp-1) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD44. The Eagle Biosciences Human CD44 ELISA Assay kit is for Research Use Only.