Human CD21 ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
C2131-K01
102994-034EA 830.38 USD
102994-034
Human CD21 ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Human CD21 ELISA determines CD21 in human serum, plasma, solutions or cell culture medium.
  • Dynamic Range: 3.12 U / ml - 100 U / ml
  • Sensitivity: 3 U / ml
  • Incubation: 1.75 hours
The Eagle Biosciences Human CD21 ELISA is to be used for the in-vitro quantitative determination of CD21 in human serum, plasma, buffered solutions or cell culture medium. The assay recognizes natural CD21. The Eagle Biosciences Human CD21 ELISA Assay kit is for Research Use Only.