Niacin Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
KIFR003
102993-714EA 1464.62 USD
102993-714
Niacin Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
Niacin is a microtiter plate test that measures the total niacin content in serum.
  • Dynamic Range: 2 - 40 µg/L
  • Sensitivity: 2 µg/L
  • Incubation: 48 hours
The Eagle Biosciences Niacin is a microtiter plate test kit based on a microbiological assay which measures the total niacin content (nicotinic acid and nicotinamide) in serum.The test kit contains all required reagents,e.g.standard,medium and microtiter plate coated with a specific microorganism, sufficient for 96 determinations including standard curves.An ELISA reader is required for evaluation of the niacin content. The Niacin Assay kit is For Research Use Only.