EDTA disodium salt solution 0.1 M/0.2N titrant

Supplier: AQUA SOLUTIONS MS

Synonyms: Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Edetic acid disodium salt dihydrate, Titriplex III, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Edetate disodium dihydrate

3540-500ML 3540-4L 3540-4LC 3540-10L 3540-1L 3540-20L
101223-290EA 28.63 USD
101223-290 101225-796 101224-316 101225-820 101224-334 101224-892
EDTA disodium salt solution 0.1 M/0.2N titrant
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
MDL Number: MFCD00150037
CAS Number: 6381-92-6

Order Now

Specification Test Results

Actual Molarity 0.1000 ± 0.0005 Molar @ 20°C
N.I.S.T Traceable Yes
Prepared to Formulation on File Yes
Appearance and Color Clear Water White Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR