Toluene ≥99.5% ACS
Supplier: AQUA SOLUTIONS MS
Synonyms:
Methacide
, Benzene, methyl
, Methylbenzol
, Toluol
, Phenylmethane
, Methylbenzene
, Methane, phenyl-
, FM approval
T7601-4L
T7601-1L
T7601-500ML
T7601-20L
T7601-4LPVC
101223-656EA
56.06
USD
101223-656
101225-286
101223-786
101224-814
101223-276
Toluene ≥99.5% ACS
Toluene
Formula:
C₆H₅CH₃ MW: 92.14 g/mol |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 UN: 1294 ADR: ,II |
Specification Test Results
Assay | 99.5% |
Color | Clear |
Form | Liquid |
Infrared Spectrum | Conforms |
Residue after Evaporation | Max. 0.003% |
Substances Darkened by Permanganate | Passes Test |
Sulfur Compounds as (S) | Max 0.003% |
Water (H₂O) | Max. 0.03% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...