Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥98.0% (dry basis)
Supplier: Thermo Fisher Scientific
J22675-A1
J22675-36
101170-164EA
144.55
USD
101170-164
101175-778
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥98.0% (dry basis)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol Boiling Pt: 219…220 °C/10 mm Melting Pt: 171…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 14,9772 |
Specification Test Results
Assay (Dry Basis) | ≥98.0% |
Identification (FTIR) | Conforms |
Loss on Drying | ≤1.5% |
pH (5%, H₂O) | 10.0 - 11.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...