Succinic acid free acid 99+%, Ultrapure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,4-Butanedioic acid

J21900-A7 J21900-A1 J21900-36
101176-046EA 86.48 USD
101176-046 101173-926 101173-004
Succinic acid free acid 99+%, Ultrapure
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235 °C
Melting Pt: 186…188 °C
Density: 1.56
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
Merck Index: 14,8869
https://us.vwr.com/stibo/web/std.lang.all/01/38/2300138.jpg

Order Now

Specification Test Results

Assay (unspecified) ≥99.0%
Water Content (Karl Fischer Titration) ≤1.0%
Identification (FTIR) Conforms
Residue on Ignition ≤0.025%
Iron (Fe) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR