Glycine 98.5-101.5% (dry basis)

Supplier: Thermo Fisher Scientific

Synonyms: Aminoacetic acidH-Gly-OHGlycocoll

J16405-A7 J16405-A1 J16405-C4
101170-932EA 1211.26 USD
101170-932 101170-930 101175-532
Glycine 98.5-101.5% (dry basis)
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Melting Pt: ∼ 245 °C (dec.)
Density: 1.595
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 14,4491
https://us.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg

Order Now

Specification Test Results

Assay (Unspecified) 98.5 - 101.5%
Assay (Unspecified) - Hydrolysable Substances Passes Test
Chloride Content ≤70 ppm
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Impurity Content Sulfate (SO₄) ≤65 ppm
Loss on Drying ≤0.2%
Heavy Metals (Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR