Sodium carbonate 1.0 N in aqueous solution
Supplier: AQUA SOLUTIONS MS
8576-1L
8576-500ML
8576-20L
8576-4L
101223-382EA
42.12
USD
101223-382
101223-336
101225-284
101224-616
Sodium carbonate 1.0 N in aqueous solution
Sodium carbonate
Certifications: Traceable to NIST (SRM 84).
Formula:
Na₂CO₃ |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 |
|
Specification Test Results
Normality | 0.9995 - 1.0005 |
Appearence and Color | Clear Water White Liquid |
Density | 1.05037 - 1.05157 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...