Potassium hydroxide 0.01 N in isopropanol (IPA) (Prepared to ASTM D3242-98)
Supplier: AQUA SOLUTIONS MS
Synonyms:
Potash caustic
, Potassium hydrate
, Caustic potash
7235-4L
7235-20L
7235-4LC
7235-500ML
7235-1L
101204-920EA
49.14
USD
101204-920
101202-894
101433-480
101203-944
101446-042
Potassium hydroxide 0.01 N in isopropanol (IPA) (Prepared to ASTM D3242-98)
Potassium hydroxide
- N.I.S.T. Traceable to SRM 84
Formula:
KOH MW: 56.11 g/mol |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 UN: 1993 ADR: ,II |
Specification Test Results
Normality | 0.0095 - 0.0105 |
Appearence | CLEAR WATER WHITE TO LIGHT HAZY |
Density | 0.78570 - 0.78641 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...