Tiger, Ceramic Resin Fiber Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-69850 804-69851 804-69852 804-69853 804-69854 804-69880 804-69881 804-69883
76184-624BX 130.36 USD
76184-624 76184-626 76184-628 76184-630 76184-632 76184-664 76184-666 76184-668 76184-670 76184-672
Tiger, Ceramic Resin Fiber Discs, Weiler®
Abrasives
Advanced ceramic grain leverages a crystalline structure that is significantly finer than standard ceramic
  • Advanced ceramic grain delivers greater stock removal with a finer scratch pattern
  • Less pressure required to achieve consistent material removal rate
  • Use with right angle grinders
  • Resin bonded
Advanced ceramic grain removes material effortlessly while a top coat grinds cool and eliminates loading and heat buildup. For those that put a premium value on getting work done faster and optimizing disc usage, Tiger Ceramic delivers.
Additional fracture points result in a more aggressive grain that self sharpens at a higher rate.