Tiger, Ceramic Combo Wheels, Weiler®
Supplier: Safety & Industrial Supplies
804-58315 804-58316
76184-232BX 326.82 USD
76184-232 76184-234
Tiger, Ceramic Combo Wheels, Weiler®
Abrasives
Self-sharpening ceramic alumina grain delivers a superior combination of life and cut rate
  • Superior life and cut on metal
Use for cutting, edge beveling, edge softening, grinding, notching, scale removal, stock removal, weld blending, and weld removal applications.
Ideal for stainless steel and other hard-to-grind metals.