High Density Twist Knot Wire Wheel, Weiler®
Supplier: Weiler
804-09799 804-08309
76183-772CT 572.97 USD
76183-772 76183-762
High Density Twist Knot Wire Wheel, Weiler®
Abrasives
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
  • Extra high density for superior performance and high aggression
  • Heavy-duty brushing action with some flexibility
  • Steel filter material
Use for cleaning, heavy burr removal, roughening surfaces, and surface conditioning applications. Removes rust, paint, oxides, and scale.