Nylox®, Crimped-Filament Wheel Brush, Weiler
Supplier: Weiler
Nylox®
804-83050 804-83070
76184-500EA 136.55 USD
76184-500 76184-498
Nylox®, Crimped-Filament Wheel Brush, Weiler
Abrasives
For maximizing brush conformability on irregular part shapes.
  • Composite metal hub for maximizing brush conformability on irregular part shape
  • With 0.04" silicon carbide bristle
  • Use for honing cutting tools and generating specific edge profiles and radii
  • Effective for improving texture characteristics on machines or ground surfaces
Good for light duty cleaning and finishing of metals, light sanding of woods and composites, and for precision deburring of component parts.