Bobcat Flat Style Flap Discs, Weiler®
Supplier: Weiler
804-50912
76183-838EA 8.92 USD
76183-838
Bobcat Flat Style Flap Discs, Weiler®
Abrasives
Used on right angle air die grinders for smooth grinding and enhanced conformability
Use for grinding, finishing, weld blending, and deburring applications. Ideal for use on steel, most metals, and stainless steel materials.
Small size allows access to confined areas.