Vortec Pro®, Abrasive Flap Discs, Weiler®
Supplier: Safety & Industrial Supplies
804-31308 804-31310 804-31313 804-31314 804-31315 804-31317 804-31333 804-31347 804-31353 804-31367 804-31409
76183-884BX 88.66 USD
76183-884 76183-886 76183-888 76183-890 76183-892 76183-894 76183-896 76183-898 76183-900 76183-902 76183-904 76183-912
Vortec Pro®, Abrasive Flap Discs, Weiler®
Abrasives
Discs provide a high cut rate and smooth grinding action.
Phenolic backing prevents disc from scratching the workpiece and offers durability and reinforcement. Use in grinding, finishing, weld blending, and deburring applications on steel, most metals, cast iron, and non-ferrous soft metal materials. Use with right angle grinders.