Diffusion and Osmosis - Small Group Learning Kit
Supplier: Aldon
IS3704-SGL
470231-558KT 100 USD
470231-558
Diffusion and Osmosis - Small Group Learning Kit
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Observe osmosis in Living Tissue.
  • Small group learning kit
  • Student copymasters and teacher guide included
Students will study the movement of water and nutrients across a cell membrane and observe osmosis in living tissue. They will then investigate the relationship between surface area and volume as it relates to cells and diffusion. They will also examine the concept of molarity and how it relates to osmotic potential and the movement of water. Students will also be able to explain how cell size and shape affect the overall rate of nutrient intake and water elimination. Students will also use plant tissue to determine the molarities of unknown solutions based on the direction and degree of water movement.