KIMTECH PURE® A8 Certified Lab Coat with Knit Collar, Kimberly Clark Professional
Supplier: Kimberly-Clark
51905
76199-148CS 181.9 USD
76199-148
KIMTECH PURE® A8 Certified Lab Coat with Knit Collar, Kimberly Clark Professional
Disposable Garments
Workers need to be protected and feel at ease in their graments. The protective three-layer spunbond-meltblown-spunbond (SMS) fabric deliver protection against light fluid contact and have soft knit cuffs for added comfort.
  • Breathable, three-layer SMS fabric
  • Soft knit cuffs for added comfort and protection
  • Recyclable through the Rightcycle Program
These lab coats feature a five-snap front closure, one breast pocket, two hip pockets, and knit collar and cuffs.