Condenser, CR Assembly, Rotary Evaporators, Chemglass
Supplier: Chemglass
CG-1335-10 CG-1335-11 CG-1333-14 CG-1333-15 CG-1333-20
80060-968EA 1845.96 USD
80060-968 80060-970 80060-956 80060-958 80060-960 80060-962
Condenser, CR Assembly, Rotary Evaporators, Chemglass
Condensers
This two-piece cold trap style condenser is for use with rotary evaporators.
The condenser has a removable hose connection using GL-14 threads and is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on the outer portion of condenser only. It has a 45/40 lower inner drip joint, a 19/38 stopcock plug.