Proto® Foot Pound/Meter Kilogram Dial Torque Wrenches, Essendant
Supplier: Stanley
577-6134F 577-6149F
76189-724EA 1146.24 USD
76189-724 76189-726
Proto® Foot Pound/Meter Kilogram Dial Torque Wrenches, Essendant
Wrenches
Memory needle pinpoints peak torque.
  • Dual scales
  • Ball locking
  • Male square drive
  • Alloy steel
  • Inch/metric
Easy to read dials. Withstands up to 20% accidental overload without losing calibration.