Proto® Spline Double Box Ratcheting Wrenches, Essendant
Supplier: Safety & Industrial Supplies
577-SBV0709 577-SBV0810 577-SBV1214 577-SBV1618 577-SBV2022 577-SBV2428 577-SBV2630 577-SBV3236
76190-070EA 42.74 USD
76190-070 76190-072 76190-074 76190-076 76190-078 76190-080 76190-082 76190-084
Proto® Spline Double Box Ratcheting Wrenches, Essendant
Wrenches
Black chrome finish is corrosion resistant and visually distinctive.
  • Box/box opening type
  • Inch measuirng system
  • Black chrome finish
Ratcheting spline box end is countersunk for easy application to the fastener. Spline design also helps prevent rounding of corners and other fastener damage that could lead to Foreign Object Damage (FOD). Spline design enables the wrench to contact conventional fasteners away from the corners, which help apply force over a large area helping to reduce risk of fastener rounding. Spline design was created for use with specialty spline bolts which are used when weight and size of bolt heads must be minimized. The fractional spline wrenches can be used on fractional 4, 6, 12 point and spline fasteners. The high contact surface area which is away from fastener points enables turning of highly rounded fasteners.