Proto® Angle Impact Wrenches, ORS Nasco
Supplier: Safety & Industrial Supplies
577-138WA 577-150WA
76189-304EA 486.58 USD
76189-304 76189-322
Proto® Angle Impact Wrenches, ORS Nasco
Wrenches
Low profile head improves access to tight areas while housing design contains gearing to reduce maintenance issues.
  • Trigger forward/reverse control
Impact mechanism provides high torque with little reaction, improving safety and reducing effort. Feather start trigger and forward/reverse ring for ease-of-use and increased productivity. Exhaust deflectors with integrated tethering connections for reduced noise and improved safety when working at heights. AJ2.