Proto® Torqueplus™ Sockets, ³/₈", Stanley® Products
Supplier: Safety & Industrial Supplies
577-5217 577-5225
76189-232EA 11.31 USD
76189-232 76189-432
Proto® Torqueplus™ Sockets, ³/₈", Stanley® Products
Wrenches
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded or rusted fasteners.
  • Square drive
  • Nickel chrome finish
  • Forged alloy steel
  • Inch measuring system
  • Ball locking
Proto Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners.