Proto® Crowfoot Wrenches, ORS Nasco
Supplier: Safety & Industrial Supplies
577-4710CF 577-4976CF 577-4996CF 577-4909MCF 577-4936CF
76188-994BX 194.08 USD
76188-994 76189-046 76189-122 76189-398 76189-400
Proto® Crowfoot Wrenches, ORS Nasco
Wrenches
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning.
  • Open opening type
  • Forged alloy steel
  • Inch measuring system
  • Ball locking type
  • Square drive
  • Nickel chrome finish
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach. Full polish finish provides corrosion resistance and easy-to-clean surfaces.