Solo Ultra 980 Nitrile Gloves, Mapa Professional®
Supplier: Mapa Professionnel
980428 980426 980421 980420 980427
32930-776CS 1005.42 USD
32930-776 32930-778 32930-780 32930-784 32930-786
Solo Ultra 980 Nitrile Gloves, Mapa Professional®
Gloves
Nitrile gloves with excellent mechanical resistance, ideal in oily environments and manufacturing of medicines
  • Textured grip offers added safety when working in wet and dry conditions
  • Exterior finish: smooth with pebbled fingertips
  • Chlorinated interior finish
Packaging: 100 gloves/box and 1000 gloves/carton.