Phenyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC13069-0010
AC13069-2500
103525-376EA
185.23
USD
103525-376
103525-380
Phenyl isocyanate 99%
Phenyl isocyanate
Formula:
C₇H₅NO MW: 119.12 g/mol Boiling Pt: 162…163 °C (760 mmHg) Melting Pt: –31.3 °C Density: 1.095 g/cm³ Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001994 CAS Number: 103-71-9 EINECS: 203-137-6 UN: 2487 ADR: 6.1,I |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5350 to 1.5380 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...