Chloroquine diphosphate salt 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloroquine phosphate7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline DiphosphateN4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine DiphosphateN4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate)

AC45524-5000 AC45524-0250 AC45524-1000
103518-910EA 53.34 USD
103518-910 103518-912 103518-200
Chloroquine diphosphate salt 98%
Chloroquine diphosphate salt
Formula: C₁₈H₂₆ClN₃
MW: 515.86 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00069852
CAS Number: 50-63-5

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105 °C, 16 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR