α-Chlorothioanisole 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Chloromethyl phenyl sulphide
, Chloromethyl phenyl sulfide
AC20236-0050
AC20236-0250
103509-644EA
74.2
USD
103509-644
103509-646
α-Chlorothioanisole 97%
α-Chlorothioanisole
Formula:
C₇H₇ClS Boiling Pt: 66 °C (.3hPa) Density: 1.23 g/cm³ Flash Pt: 101 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000921 CAS Number: 7205-91-6 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...