Carbenicillin disodium salt 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2NaNSC 111071α-Carboxybenzylpenicillin disodium saltSodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

AC45536-0010 AC45536-0050
103517-608EA 123.25 USD
103517-608 103517-610
Carbenicillin disodium salt 90%
Carbenicillin disodium salt
Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Boiling Pt: 738 °C
Melting Pt: >200 °C
Flash Pt: 400 °C
Storage Temperature: Freezer
CAS Number: 4800-94-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 %
Solubility (5 % in water) Clear to light yellow
HPLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR