3'-Azido-3'-deoxythymidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Azidothymidine
, 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine
, AZT
, Zidovudine
AC45498-0010
AC45498-0050
103517-572EA
103
USD
103517-572
103517-570
3'-Azido-3'-deoxythymidine 98%
3'-Azido-3'-deoxythymidine
Formula:
C₁₀H₁₃N₅O₄ MW: 267.25 g/mol Melting Pt: 118…120 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006536 CAS Number: 30516-87-1 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 122 to 126 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...