(±)-1,2-Propanediol ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Propylene glycol(±)-1,2-Propylene glycol1,2-Dihydroxypropane1,2-Propanediol(±)-1,2-Dihydroxy propane

AC44741-5000 AC44741-0025 AC44741-1000
103516-884EA 38.02 USD
103516-884 103516-886 103516-882
(±)-1,2-Propanediol ACS
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –60 °C
Density: 1.03 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
https://us.vwr.com/stibo/web/std.lang.all/05/57/710557.jpg

Order Now

Specification Test Results

Appearance Clear viscous liquid
Water ≤0.2 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after ignition ≤0.005 %
Chloride (Cl) ≤1 ppm
Titrable acid ≤0.0005 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR