(Trimethylsilyl)diazomethane ∼2 mol/l in diethyl ether, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylsilyldiazomethane
, TMS-Diazomethane
AC42920-1000
103524-348EA
567.14
USD
103524-348
(Trimethylsilyl)diazomethane ∼2 mol/l in diethyl ether, AcroSeal™
(Trimethylsilyl)diazomethane
Formula:
C₄H₁₀N₂Si MW: 114.22 Density: 0.77 g/cm³ Flash Pt: –35 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053946 CAS Number: 18107-18-1 UN: 1992 ADR: 3,I |
Specification Test Results
Appearance | Yellow Solution (May develop some turbidity or precipitate) |
Strength | 1.9 to 2.2 M |
Strength | 0.5 % (typically) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...