Sterile Stainless Steel Surgical Blades, Propper Manufacturing
Supplier: Propper
12301000 12301100 12301200 12301500 12302000 12302200 12302400
76044-646BX 102.25 USD
76044-646 76044-648 76044-650 76044-652 76044-654 76044-656 76044-658
Sterile Stainless Steel Surgical Blades, Propper Manufacturing
Scalpels and Scalpel Assemblies Scalpel Blades
Propper sterile stainless steel surgical blades are available in a variety of precise shapes and sizes. Each sterile blade is enclosed in a heat sealed, peel pack foil envelope.
  • Superior cutting ability of stainless steel
  • Sterile blades
  • Enclosed in heat-sealed peel-pack
Description of use of each size
#10: Curved cutting edge is one of the more traditional blade shapes, making small incisions to the skin and muscle clean and precise.
#11: Elongated triangular blade with a strong tip, ideal for stab incisions. Most commonly used for the incisions of chest drains, and opening various coronary arteries.
#15: Small curved cutting edge is the most popular blade for short and precise incisions.
#20: A larger version of the No. 10 blade equipped with a curved cutting edge, used mostly in orthopedic and general surgical procedures.
#22: A tad bigger than the number #21 blade, used for skin incisions in both cardiac and thoracic surgery.
#24: Semi-circular shape, sharpened along its leading edge. Used for making long incisions in general surgery and also in autopsy procedures.