CSM Powder, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
114500012 114500022
76019-996EA 163.81 USD
76019-996 76019-998
CSM Powder, MP Biomedicals
Cell Culture Media
Complete Supplement Mixture (CSM) is a culture growth media supplement for virtually all strains of S. cerevisiae.
  • Molecular Biology Certified Yeast Growth Medium
  • Vigorous cell growth
Complete Supplement Mixture (CSM) is a culture growth media supplement for virtually all strains of S
Complete Supplement Mixture (CSM) is a time-tested dropout supplement for virtually all strains of S. cerevisiae. Cells grow vigorously in CSM supplemented medium which is easily made by adding the CSM powder to DOB of DOBA Base Medium and autoclaving.
Add 0.79g of CSM to each liter of DOB or DOBA and autoclave at 121 °C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.(Lit.)
cerevisiae.