100 and 130 Series Multi-Purpose Grease, Lubriplate®
Supplier: Lubriplate
293-L0037-035
300039-538EA 398.02 USD
300039-538
100 and 130 Series Multi-Purpose Grease, Lubriplate®
Lubricants
Designed for plain bearings, cams, slides, and other light-duty applications
  • Calcium type grease
  • 190 °F drop point (ASTM)
  • 300 SUS @ 100 °F
  • Specific gravity: 0.89 – 0.93
  • NLGI grade 3
For applications where temperatures do not exceed 170 °F/77 °C.