X-Gal Staining Kit, OZ Biosciences
Supplier: OZ Biosciences
GX10003
103258-324EA 242.08 USD
103258-324
X-Gal Staining Kit, OZ Biosciences
Assays Cellular Assays
OZ Biosciences X-Gal Staining Kit allows to visualize -gal expression through hydrolysis of the X-Gal (5-bromo-4-chloro-3-indoyl-beta-D-Galactopyranoside) substrate yielding blue precipitates
  • Convenient for all LacZ transfection assays
  • Suitable for cultured cells and tissues
  • Complete kit provided with substrate and buffers
  • Economical
LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily tested.
Delivery information: Shipped with a gel pack
Packaging: Store at +4°C except 25X Stock Solution of X-Gal -20°C