10X PBD
Supplier: MP Biomedicals
11RIST1370
76018-086EA 254.96 USD
76018-086
10X PBD
Buffers Electrophoresis Buffers Hybridization Buffers
10X Phosphate-Buffered Detergent.
  • Enzymatic or fluorescence in situ hybridization
  • Nucleic Acid Analysis
  • Formulation: SSC 4X and 0.5% TWIN 20
10X Phosphate buffered detergent (PBD) used in enzymatic or fluorescence in situ hybridization (FISH). Intended for use with paraffin embedded, formalin-fixed tissue sections, whole cells interphase nuclei, and metaphase chromosomes.