Agarose, white powder, high resolution for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

11AGAR0050
76180-732EA 433.75 USD
76180-732
Agarose, white powder, high resolution for molecular biology
Agarose

Agarose High Resolution is an intermediate melting agarose with approximately twice the resolution capability of routine agarose. It is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae. It is ideal for the resolution of small DNA fragments in the range 20 bp to 2000 bp.
Agarose high resolution is an intermediate melting and gelling point agarose which exhibits superior sieving characteristics. It is recommended for analytical gels of DNA fragments under 1000 bp and is ideal for small DNA fragments down to 20 bp can be separated on 6 % gels. It resolves fragments differing in size by only 2 bp. Its use avoids the need for PAGE Southern and Northern transfers can be achieved efficiently from them at concentrations up to 2%. DNA fragments can be purified with high efficiency from this agarose gels up to 4% by using either a purification kit or an electroelution system.


  • Molecular Biology Grade
  • Suitable for electrophoresis of nucleic acids
  • Presentation: White Powder
  • Soluble in water (15 mg/mL)
  • Store below 22-25 °C.

Melting Pt: < 67 °C (1.5% gel)
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
https://us.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg

Order Now

Specification Test Results

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR