X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by TLC), white powder, Ultrapure
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
BCIG
, X-GAL
, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
114063102
76020-284EA
212.02
USD
76020-284
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by TLC), white powder, Ultrapure
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
- Ultra Pure Grade
- Purity: (TLC) >99%
- Optical Rotation: -61.9%(c=1, DMF:water=1:1, Lit.)
- Presentation: White Powder
- Soluble in dimethylformamide (2% in DMF), Dimethyl sulfoxide (20 mg/mL) & DMF:MeOH (1:1) (50 mg/mL)
- Storage temperature: -20°C
Formula:
C₁₄H₁₅BrClNO₆ MW: 408.63 g/mol |
MDL Number:
MFCD00005666 CAS Number: 7240-90-6 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...