ZING Green Safety Eco Safety Sign BIOHAZARD Peligro Biologico
Supplier: ZING Enterprises
10056 10056A 10056S 20056 20056A 20056S
76185-690EA 24.68 USD
76185-690 76185-692 76185-694 76185-696 76185-698 76185-700
ZING Green Safety Eco Safety Sign BIOHAZARD Peligro Biologico
Signs
The sign is available in 2 sizes, 10 x 7", and 14 x 10", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.