ZING Green Safety Eco Safety Sign NOTICE, No Food or Drink in This Area
Supplier: ZING Green Safety Products
2991A 1991S 1991A 2991 2991S 1991
76185-334EA 30.44 USD
76185-334 76185-340 76185-164 76185-342 76185-336 76185-338
ZING Green Safety Eco Safety Sign NOTICE, No Food or Drink in This Area
Signs
The sign is available in 2 sizes, 7 x 10", and 10 x 14", and three eco-friendly formats.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks
Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting.
Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure.
All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.