Ergonomic Tool Holder, MSA
Supplier: MSA
454-507005
300009-013EA 122.11 USD
300009-013
Ergonomic Tool Holder, MSA
Tools Storage
Use with wrenches, chisels, or any tools up to 18" in length.
  • Vibration-absorbing design
  • Attach to broom handles and mops for better grip
  • Eases awkward hand positioning
  • 2.75" opening size