Human iPSC-Derived Motor Neurons, iXCells Biotechnologies
Supplier: IXCELLS BIOTECHNOLOGIES USA MS
Human iPSC-Derived Motor Neurons; Highly pure motor neurons with functional validation.
- High-quality human motor neurons derived from iPSCs
- High purity
- Formation of neuromuscular junction when co-cultured with myotubes
- IRB-approved
Human iPSC-derived motor neuron; Frozen; iXCells™ hiPSC-derived motor neurons express typical markers HB9 (MNX1), ISL1, CHAT, with the purity higher than 85%. Functional validation of iPSC-derived motor neurons has been done with neuromuscular junction formation. Available for cryopreserved and live culture on plate formats. Negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.