Fluorophore-Infiltrated Resin Microscopy (FIRM), Electron Microscopy Science
Supplier: Electron Microscopy Sciences
12780-05 12780-06
103303-368EA 172.65 USD
103303-368 103303-370
Fluorophore-Infiltrated Resin Microscopy (FIRM), Electron Microscopy Science
Stains Microbiology Stains
Rapid staining technique that provides high-contrast fluorescence microscopy imaging of resin embedded samples, reminiscint of low magnification transmission electron microscopy.
  • Works with Epoxy and Acrylic type resins
  • Perfect for previewing specimens prior to EM Analysis
  • High Throughput, high contrast image phenotyping of model organisms such as drosophila
  • Ideal for material and food science analytical microscopy
This rapid staining technique provides high-contrast fluorescence microscopy imaging of resin embedded samples, reminiscint of low magnification transmission electron microscopy. FIRM staining is unique in that the brightly fluorescent dye infiltrates the resin, revealing tissue structures in negative relief, with extremely high contrast and resolution.
To use simply mount your sections on glass slides, air dry, and overlay with 100 microliters FIRM for 30 seconds, wash and coverslip with water or an aqueous mountant. View in Rhodamine channel of a standard wide field Fluorescence Microscope.
To use simply mount your sections on glass slides, air dry, and overlay with 100 microliters FIRM for 30 seconds, wash and coverslip with water or an aqueous mountant. View in Rhodamine channel of a standard wide field Fluorescence Microscope.
Perfect for high throughput, high contrast image phenotyping of model organisms such as drosophila.