Advantage® 420 Series Reusable Half Mask Respirators, MSA
Supplier: Safety & Industrial Supplies
454-10102182 454-10102184
300081-010EA 50.79 USD
300081-010 300081-014
Advantage® 420 Series Reusable Half Mask Respirators, MSA
Respirators Reusable Respirators Half Face Respirators
With its UniBond over-mold facepiece, AnthroCurve™ face seal, and low profile design, the Advantage® 420 Respirator is the superior option for respiratory protection.
The over-mold facepiece and face seal ensure the unit provides an excellent fit and comfort to the wearer. The unique yoke and harness design (patent pending) allows a user to easily don and doff the state-of-the-art respirator while maintaining a customized fit. Other features include a ComforTop headband, a built-in moisture release chin cup hole, and the extra-wide head and neck straps.
Delivery information: More Product Information